missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-phenylhexane, 98+%, Thermo Scientific™

Product Code. 11342796
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342796 250 mg 250mg
11332796 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342796 Supplier Thermo Scientific Alfa Aesar Supplier No. L08949.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71434-68-9
Molecular Formula C12H17Cl
Molecular Weight (g/mol) 196.72
MDL Number MFCD00013698
InChI Key RCVNHNOEUMFFDY-UHFFFAOYSA-N
Synonym 1-chloro-6-phenylhexane, 6-chlorohexyl benzene, 6-phenylhexylchloride, 6-phenylhexyl chloride, 6-phenyl-1-hexylchloride, 6-chlorohexyl benzene #
PubChem CID 522411
IUPAC Name 6-chlorohexylbenzene
SMILES ClCCCCCCC1=CC=CC=C1

Specifications

Boiling Point 88°C to 91°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5105
Quantity 250 mg
Beilstein 2088276
Formula Weight 196.72
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-6-phenylhexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.