accessibility menu, dialog, popup

UserName

HOBOHOH

CAS RN 71597-85-8

IUPAC Name: (4-hydroxyphenyl)boronic acid
Synonyms: 4-hydroxyphenylboronic acid
Molecular Weight (g/mol): 137.93
Molecular Formula: C6H7BO3
InChi Key: COIQUVGFTILYGA-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(O)C=C1
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results