accessibility menu, dialog, popup

UserName

CH3H3CCH3H3CCH3CH3

CAS RN 717-74-8

IUPAC Name: 1,3,5-tris(propan-2-yl)benzene
Synonyms: 1,3,5-triisopropylbenzene
Molecular Weight (g/mol): 204.36
Molecular Formula: C15H24
InChi Key: VUMCUSHVMYIRMB-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)