accessibility menu, dialog, popup

UserName

CAS RN 717-94-2

H3COOOOH3C

CAS RN 717-94-2

IUPAC Name: 3-(3,5-dimethoxyphenyl)propanoate
Synonyms: 3-(3,5-dimethoxyphenyl)propanoate
Molecular Weight (g/mol): 209.22
Molecular Formula: C11H13O4
InChi Key: LMBOJOXVLORKSQ-UHFFFAOYSA-M
SMILES: COC1=CC(OC)=CC(CCC([O-])=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results