accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 7194-86-7

IUPAC Name: nonatriacontane
Synonyms: nonatriacontane
Molecular Weight (g/mol): 549.07
Molecular Formula: C39H80
InChi Key: SNXOSZZZNFRFNZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results