missing translation for 'onlineSavingsMsg'
Learn More

Nonatriacontane, TRC

Product Code. 30466398
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30466398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466398 Supplier Toronto Research Chemicals Supplier No. TRCN458450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7194-86-7
Molecular Formula C39 H80
Molecular Weight (g/mol) 549.05
Synonym Nonatriacontane, n-Nonatriacontane
IUPAC Name nonatriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C39H80/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-39H2,1-2H3
Formula Weight 548.626
Chemical Name or Material n-Nonatriacontane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.