accessibility menu, dialog, popup

UserName

CAS RN 72235-55-3

H2NClF

CAS RN 72235-55-3

IUPAC Name: 1-(3-chloro-2-fluorophenyl)methanamine
Synonyms: 1-(3-chloro-2-fluorophenyl)methanamine
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: LYKWZKBLNOCJMA-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results