CAS RN 72646-00-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(3-Chloro-2-methylphenyl)-2-hydroxynicotinamide, TRC
CAS: 72646-00-5 Molecular Formula: C13 H11 Cl N2 O2 Molecular Weight (g/mol): 262.69 Synonym: N-(3-Chloro-2-methylphenyl)-1,2-dihydro-2-oxo-3-pyridinecarboxamide,N-(3-Chloro-2-methylphenyl)-2-oxo-1,2-dihydropyridine-3-carboxamide IUPAC Name: N-(3-chloro-2-methylphenyl)-2-hydroxypyridine-3-carboxamide SMILES: Cc1c(Cl)cccc1NC(=O)c2cccnc2O