accessibility menu, dialog, popup

UserName

CAS RN 727733-92-8

NN

CAS RN 727733-92-8

IUPAC Name: 4-[(piperidin-1-yl)methyl]benzonitrile
Synonyms: 4-(piperidin-1-ylmethyl)benzonitrile
Molecular Weight (g/mol): 200.29
Molecular Formula: C13H16N2
InChi Key: JOGVBKNLRQCYKS-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(CN2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results