missing translation for 'onlineSavingsMsg'
Learn More

4-(Piperidin-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10371483
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10371483 1 g 1g
10066674 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371483 Supplier Thermo Scientific Maybridge Supplier No. CC45516DA
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 727733-92-8
Molecular Formula C13H16N2
Molecular Weight (g/mol) 200.285
MDL Number MFCD06796182
InChI Key JOGVBKNLRQCYKS-UHFFFAOYSA-N
Synonym 4-piperidin-1-ylmethyl benzonitrile, 1-4-cyanobenzyl piperidine, 4-piperidinomethyl benzonitrile, 4-piperidylmethyl benzenecarbonitrile, 4-piperidin-1-ylmethyl-benzonitrile, benzonitrile,4-1-piperidinylmethyl
PubChem CID 7537603
IUPAC Name 4-(piperidin-1-ylmethyl)benzonitrile
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.