CAS RN 7284-16-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Nitrophenyl N,N-Diacetyl-Beta-D-chitobioside, TRC
CAS: 7284-16-4 Molecular Formula: C22 H31 N3 O13 Molecular Weight (g/mol): 545.49 IUPAC Name: N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4R,5R,6S)-5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide SMILES: CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@H]2[C@H](O)[C@@H](NC(=O)C)[C@H](Oc3ccc(cc3)[N+](=O)[O-])O[C@@H]2CO