CAS RN 7298-94-4
CAS RN 7298-94-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
a-Nicotinamide Mononucleotide (>85%), TRC
CAS: 7298-94-4 Molecular Formula: C11H15N2O8P Molecular Weight (g/mol): 334.22 Synonym: alpha-NMN,3-(Aminocarbonyl)-1-(5-O-phosphono-alpha-D-ribofuranosyl)pyridinium,((2R,3S,4R,5S)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl Hydrogen Phosphate IUPAC Name: [(2R,3S,4R,5S)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate SMILES: NC(=O)c1ccc[n+](c1)[C@H]2O[C@H](COP(=O)(O)[O-])[C@@H](O)[C@H]2O