missing translation for 'onlineSavingsMsg'
Learn More

a-Nicotinamide Mononucleotide (>85%), TRC

Product Code. 30424543
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424543 5 mg -
30413637 1 mg 1mg
30410439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424543 Supplier Toronto Research Chemicals Supplier No. TRCN7512705MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7298-94-4
Molecular Formula C11H15N2O8P
Molecular Weight (g/mol) 334.22
Synonym alpha-NMN, 3-(Aminocarbonyl)-1-(5-O-phosphono-alpha-D-ribofuranosyl)pyridinium, ((2R,3S,4R,5S)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl Hydrogen Phosphate
IUPAC Name [(2R,3S,4R,5S)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
SMILES NC(=O)c1ccc[n+](c1)[C@H]2O[C@H](COP(=O)(O)[O-])[C@@H](O)[C@H]2O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H15N2O8P/c12-10(16)6-2-1-3-13(4-6)11-9(15)8(14)7(21-11)5-20-22(17,18)19/h1-4,7-9,11,14-15H,5H2,(H3-,12,16,17,18,19)/t7-,8-,9-,11+/m1/s1
Formula Weight 334.0566
Chemical Name or Material alpha-Nicotinamide Mononucleotide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.