accessibility menu, dialog, popup

UserName

BrBrP

CAS RN 7301-93-1

IUPAC Name: (2-bromoethyl)triphenylphosphanium bromide
Synonyms: (2-bromoethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 450.15
Molecular Formula: C20H19Br2P
InChi Key: ZHLVWYZWQPNQDQ-UHFFFAOYSA-M
SMILES: [Br-].BrCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results