accessibility menu, dialog, popup

UserName

CAS RN 73096-39-6

OHONNHNN

CAS RN 73096-39-6

IUPAC Name: 3-(2H-1,2,3,4-tetrazol-5-yl)benzoic acid
Synonyms: 3-(2H-1,2,3,4-tetrazol-5-yl)benzoic acid
Molecular Weight (g/mol): 190.16
Molecular Formula: C8H6N4O2
InChi Key: MIERACSHCALJOM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=NNN=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results