accessibility menu, dialog, popup

UserName

PBr

CAS RN 7333-52-0

IUPAC Name: cyclopentyltriphenylphosphanium bromide
Synonyms: cyclopentyltriphenylphosphanium bromide
Molecular Weight (g/mol): 411.32
Molecular Formula: C23H24BrP
InChi Key: WZYWSVSFFTZZPE-UHFFFAOYSA-M
SMILES: [Br-].C1CCC(C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results