CAS RN 73356-19-1
CAS RN 73356-19-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-(N-Methylaminomethyl)anthracene, TRC
CAS: 73356-19-1 Molecular Formula: C16H15N Molecular Weight (g/mol): 221.3 Synonym: N-Methyl-9-anthracenemethylamine,9-Methylaminomethylanthracene IUPAC Name: 1-anthracen-9-yl-N-methylmethanamine SMILES: CNCC1=C(C=CC=C2)C2=CC3=C1C=CC=C3