missing translation for 'onlineSavingsMsg'
Learn More

9-(N-Methylaminomethyl)anthracene, TRC

Product Code. 30483722
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483722 100 mg 100mg
30481302 1 g 1g
30482846 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483722 Supplier Toronto Research Chemicals Supplier No. TRCM288080100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73356-19-1
Molecular Formula C16H15N
Molecular Weight (g/mol) 221.3
Synonym N-Methyl-9-anthracenemethylamine, 9-Methylaminomethylanthracene
IUPAC Name 1-anthracen-9-yl-N-methylmethanamine
SMILES CNCC1=C(C=CC=C2)C2=CC3=C1C=CC=C3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H15N/c1-17-11-16-14-8-4-2-6-12(14)10-13-7-3-5-9-15(13)16/h2-10,17H,11H2,1H3
Formula Weight 221.1204
Chemical Name or Material 9-(N-Methylaminomethyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.