CAS RN 73499-12-4
CAS RN 73499-12-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cortisol 21-m-Maleimidobenzoate, TRC
CAS: 73499-12-4 Molecular Formula: C32H35NO8 Molecular Weight (g/mol): 561.62 Synonym: 2-((8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate IUPAC Name: [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 3-(2,5-dioxopyrrol-1-yl)benzoate SMILES: C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)c5cccc(c5)N6C(=O)C=CC6=O)[C@@]4(C)C[C@H](O)[C@H]23