missing translation for 'onlineSavingsMsg'
Learn More

Cortisol 21-m-Maleimidobenzoate, TRC

Product Code. 30366508
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366508 25 mg -
30351007 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30366508 Supplier Toronto Research Chemicals Supplier No. TRCC69637025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73499-12-4
Molecular Formula C32H35NO8
Molecular Weight (g/mol) 561.62
Synonym 2-((8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 3-(2,5-dioxopyrrol-1-yl)benzoate
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)c5cccc(c5)N6C(=O)C=CC6=O)[C@@]4(C)C[C@H](O)[C@H]23

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C32H35NO8/c1-30-12-10-21(34)15-19(30)6-7-22-23-11-13-32(40,31(23,2)16-24(35)28(22)30)25(36)17-41-29(39)18-4-3-5-20(14-18)33-26(37)8-9-27(33)38/h3-5,8-9,14-15,22-24,28,35,40H,6-7,10-13,16-17H2,1-2H3/t22-,23-,24-,28+,30-,31-,32-/m0/s1
Formula Weight 561.2363
Chemical Name or Material Cortisol 21-m-Maleimidobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.