accessibility menu, dialog, popup

UserName

CH3CH3OOHO

CAS RN 7356-41-4

IUPAC Name: 2-(2,5-dimethylphenoxy)acetic acid
Synonyms: 2,5-dimethylphenoxyacetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: RSJMMLSDGNQOEO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(OCC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results