CAS RN 7372-88-5
CAS RN 7372-88-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methyldibenzothiophene, TRC
CAS: 7372-88-5 Molecular Formula: C13 H10 S Molecular Weight (g/mol): 198.28 Synonym: 4-MDBT,4-Methyldibenzo[b,d]thiophene IUPAC Name: 4-methyldibenzothiophene SMILES: Cc1cccc2c3ccccc3sc12