missing translation for 'onlineSavingsMsg'
Learn More

4-Methyldibenzothiophene, TRC

Product Code. 17942135
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942135 500 mg 500mg
17901405 1 g 1g
17933015 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942135 Supplier Toronto Research Chemicals Supplier No. TRCM299765500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7372-88-5
Molecular Formula C13 H10 S
Molecular Weight (g/mol) 198.28
Synonym 4-MDBT, 4-Methyldibenzo[b,d]thiophene
IUPAC Name 4-methyldibenzothiophene
SMILES Cc1cccc2c3ccccc3sc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H10S/c1-9-5-4-7-11-10-6-2-3-8-12(10)14-13(9)11/h2-8H,1H3
Formula Weight 198.0503
Percent Purity >95%
Chemical Name or Material 4-Methyldibenzothiophene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.