accessibility menu, dialog, popup

UserName

CAS RN 7383-77-9

IUPAC Name: dimethyl(pent-2-yn-1-yl)amine
Synonyms: dimethyl(pent-2-yn-1-yl)amine
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: XOJYPYPTLZUHAG-UHFFFAOYSA-N
SMILES: CCC#CCN(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results