accessibility menu, dialog, popup

UserName

BClClHOOH

CAS RN 73852-17-2

IUPAC Name: (2,6-dichlorophenyl)boronic acid
Synonyms: 2,6-dichlorophenylboronic acid
Molecular Weight (g/mol): 190.81
Molecular Formula: C6H5BCl2O2
InChi Key: CXDPUSMFYPQXCV-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)