missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15439109
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439109 250 mg 250mg
11486870 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439109 Supplier Thermo Scientific Alfa Aesar Supplier No. L19816.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73852-17-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00064869
InChI Key CXDPUSMFYPQXCV-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid
PubChem CID 2734332
IUPAC Name (2,6-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 151°C to 152°C
Quantity 250 mg
Beilstein 4802506
Solubility Information Insoluble in water.
Formula Weight 190.82
Percent Purity 95%
Chemical Name or Material 2,6-Dichlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.