accessibility menu, dialog, popup

UserName

H3CCH3ONHOHNO

CAS RN 7391-69-7

IUPAC Name: 5-(propan-2-yl)-1,3-diazinane-2,4,6-trione
Synonyms: barbituric acid, 5-ipr
Molecular Weight (g/mol): 170.17
Molecular Formula: C7H10N2O3
InChi Key: GFPIGNBQTXNNAG-UHFFFAOYSA-N
SMILES: CC(C)C1C(=O)NC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results