missing translation for 'onlineSavingsMsg'
Learn More

Isopropylbarbituric acid, 97%, Thermo Scientific™

Product Code. 15296028 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15296028 5 g 5g
15206038 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15296028 Supplier Thermo Scientific Acros Supplier No. 452670050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7391-69-7
Molecular Formula C7H10N2O3
Molecular Weight (g/mol) 170.168
InChI Key GFPIGNBQTXNNAG-UHFFFAOYSA-N
Synonym 5-isopropylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-isopropylpyrimidine-2,4,6 1h,3h,5h-trione, 5-isopropyl barbituric acid, barbituric acid, 5-isopropyl, barbituric acid, 5-ipr, 2,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-1-methylethyl pyrimidine-2,4,6 1h,3h,5h-trione
PubChem CID 81874
IUPAC Name 5-propan-2-yl-1,3-diazinane-2,4,6-trione
SMILES CC(C)C1C(=O)NC(=O)NC1=O

Specifications

Melting Point 215.0°C to 217.0°C
Formula Weight 170.17
Quantity 5 g
Chemical Name or Material Isopropylbarbituric acid
compliance-icons compliance-icons
Product Identifier
  • Isopropylbarbituric acid, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.