accessibility menu, dialog, popup

UserName

CAS RN 73948-18-2

H3CSClOO

CAS RN 73948-18-2

IUPAC Name: 4-pentylbenzene-1-sulfonyl chloride
Synonyms: 4-pentylbenzenesulfonyl chloride
Molecular Weight (g/mol): 246.75
Molecular Formula: C11H15ClO2S
InChi Key: XDWXZRVWJHEWMT-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)