missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzenesulfonyl chloride, 97%

Product Code. 11438777
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438777 10 g 10g
11428777 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438777 Supplier Thermo Scientific Alfa Aesar Supplier No. A19929.09
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 73948-18-2 | C11H15ClO2S | 246.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73948-18-2
Molecular Formula C11H15ClO2S
Molecular Weight (g/mol) 246.75
MDL Number MFCD00173759
InChI Key XDWXZRVWJHEWMT-UHFFFAOYSA-N
Synonym 4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
PubChem CID 144625
IUPAC Name 4-pentylbenzenesulfonyl chloride
SMILES CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 39°C to 44°C
Quantity 10 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 246.76
Percent Purity 97%
Chemical Name or Material 4-n-Pentylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.