accessibility menu, dialog, popup

UserName

CH3OOOOCH3

CAS RN 74079-01-9

IUPAC Name: 1,4-dimethyl benzene-1,4-dicarboxylate
Synonyms: dimethyl terephthalate dimethylterephthalate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: WOZVHXUHUFLZGK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results