CAS RN 74288-40-7
CAS RN 74288-40-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
p-Nitrobenzyl 6-(1’-Hydroxyethyl)-azabicyclo(3.2.0)heptane-3,7-dione-2-carboxylate, TRC
CAS: 74288-40-7 Molecular Formula: C16 H16 N2 O7 Molecular Weight (g/mol): 348.31 IUPAC Name: (4-nitrophenyl)methyl (5R,6S)-6-[(1R)-1-hydroxyethyl]-3,7-dioxo-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: C[C@@H](O)[C@@H]1[C@H]2CC(=O)C(N2C1=O)C(=O)OCc3ccc(cc3)[N+](=O)[O-]