accessibility menu, dialog, popup

UserName

CH3S

CAS RN 7433-79-6

IUPAC Name: 2-(methylsulfanyl)naphthalene
Synonyms: 2-(methylsulfanyl)naphthalene
Molecular Weight (g/mol): 174.26
Molecular Formula: C11H10S
InChi Key: SSKUUDUKJMIOKQ-UHFFFAOYSA-N
SMILES: CSC1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results