accessibility menu, dialog, popup

UserName

U

CAS RN 7440-61-1

IUPAC Name: uranium
Synonyms: uranium Uran uranium(0) uranium atom uranio
Molecular Weight (g/mol): 238.03
Molecular Formula: U
InChi Key: JFALSRSLKYAFGM-UHFFFAOYSA-N
SMILES: [U]
Solvent or Matrix 
  • (8)
Concentration 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results