accessibility menu, dialog, popup

UserName

H3COBr

CAS RN 74447-73-7

IUPAC Name: 3-bromo-4-methoxy-1,1'-biphenyl
Synonyms: 3-bromo-4-methoxy-1,1'-biphenyl
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: QJGJVVIDRMVQMR-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1Br)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results