accessibility menu, dialog, popup

UserName

ClOHN

CAS RN 7461-34-9

IUPAC Name: N-benzyl-4-chlorobenzamide
Synonyms: N-benzyl-4-chlorobenzamide
Molecular Weight (g/mol): 245.71
Molecular Formula: C14H12ClNO
InChi Key: LSMWDKIFKGLNSW-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results