accessibility menu, dialog, popup

UserName

CAS RN 7468-67-9

ON

CAS RN 7468-67-9

IUPAC Name: 2-formylbenzonitrile
Synonyms: 2-formylbenzonitrile
Molecular Weight (g/mol): 131.13
Molecular Formula: C8H5NO
InChi Key: QVTPWONEVZJCCS-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1C#N
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)