accessibility menu, dialog, popup

UserName

CH3OHNH3C

CAS RN 74786-81-5

IUPAC Name: N,3-dimethylbenzamide
Synonyms: N,3-dimethylbenzamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: WKSODUAHKWBHQZ-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC(C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results