accessibility menu, dialog, popup

UserName

CH3AsCH3OHO

CAS RN 75-60-5

IUPAC Name: dimethylarsinic acid
Synonyms: ansar caswell no. 133 acide cacodylique dimethylarsenic kakodylsaeure erase arsinic acid, as,as-dimethyl- Me2As(=O)OH acid, cacodylic sylvicor
Molecular Weight (g/mol): 138
Molecular Formula: C2H7AsO2
InChi Key: OGGXGZAMXPVRFZ-UHFFFAOYSA-N
SMILES: C[As](C)(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)