missing translation for 'onlineSavingsMsg'
Learn More

Cacodylic acid, 98%

Product Code. 10770785 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10770785 5 g Glass bottle 5g
10711551 10 g Glass bottle 10g
15739128 50 g Glass bottle 50g
10701355 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10770785 Supplier Thermo Scientific Acros Supplier No. 318150050
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 75-60-5 | C2H7AsO2 | 138.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-60-5
Molecular Formula C2H7AsO2
Molecular Weight (g/mol) 138.00
MDL Number MFCD00002095
InChI Key OGGXGZAMXPVRFZ-UHFFFAOYSA-N
Synonym cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510
PubChem CID 2513
ChEBI CHEBI:48765
IUPAC Name dimethylarsinic acid
SMILES C[As](C)(O)=O

Specifications

Melting Point 195°C to 198°C
Color White
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Linear Formula (CH3)2As(O)OH
Merck Index 15, 1609
Formula Weight 138.01
Percent Purity ≥97.5%
Chemical Name or Material Cacodylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.