CAS RN 7534-94-3
CAS RN 7534-94-3
IUPAC Name:
1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl 2-methylprop-2-enoate
Synonyms:
isobornyl methacrylate
Molecular Weight (g/mol):
222.33
Molecular Formula:
C14H22O2
InChi Key:
IAXXETNIOYFMLW-UHFFFAOYNA-N
SMILES:
CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C
1
–
1
of
1
results
1
Promotions Available
Isobornyl methacrylate, 85-90%, stabilized
Isobornyl methacrylate, 85-90%, stabilized
CAS: 7534-94-3 Molecular Formula: C14H22O2 Molecular Weight (g/mol): 222.33 MDL Number: MFCD00081070 InChI Key: IAXXETNIOYFMLW-UHFFFAOYNA-N Synonym: ibma,isobornyl methacrylate,methacrylic acid isobornyl ester,iso-bornyl methacrylate,isobornyl methacrylate, technical grade,isobornyl methacrylate, stabilized with mehq,exo-1,7,7-trimethylbicyclo 2.2.1 hept-,2-yl methacrylate,1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate PubChem CID: 71311141 IUPAC Name: [(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C