missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl methacrylate, 85-90%, stabilized

Product Code. 10239203 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10239203

Brand: Thermo Scientific Acros 292090010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7534-94-3 | C14H22O2 | 222.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7534-94-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00081070
InChI Key IAXXETNIOYFMLW-UHFFFAOYNA-N
Synonym ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate
PubChem CID 71311141
IUPAC Name [(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C

Specifications

Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 245.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780
Quantity 1 kg
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Stabilizer 100 to 200ppm MEHQ
Formula Weight 222.33
Percent Purity 85-90%
Physical Form Liquid
Chemical Name or Material Isobornyl methacrylate, 85-90%
Show More Show Less
compliance-icons
Product Identifier
  • Isobornyl methacrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.