accessibility menu, dialog, popup

UserName

OHFFFFFFFFFFFFFFFFHO

CAS RN 754-96-1

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol
Molecular Weight (g/mol): 462.13
Molecular Formula: C10H6F16O2
InChi Key: NSKCTPBWPZPFHW-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results