missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,10H,10H-Perfluoro-1,10-decanediol, 96%, Thermo Scientific Chemicals

Product Code. 11313776
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313776 5 g 5g
11303776 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313776 Supplier Thermo Scientific Alfa Aesar Supplier No. L16823.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-96-1
Molecular Formula C10H6F16O2
Molecular Weight (g/mol) 462.13
MDL Number MFCD00192200
InChI Key NSKCTPBWPZPFHW-UHFFFAOYSA-N
PubChem CID 2733404
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol
SMILES C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 132°C to 134°C
Boiling Point 132°C (4 mmHg)
Quantity 5 g
Beilstein 1894671
Synonym 1h,1h,10h,10h-perfluorodecane-1,10-diol, 1h,1h,10h,10h-perfluoro-1,10-decanediol, 1h,1h,10h,10h-hexadecafluoro-1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro-1,10-decanediol, 1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, hexadecafluorooctamethylenebismethanol, 1,10-decanediol,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecakis fluoranyl decane-1,10-diol
Formula Weight 462.13
Percent Purity 96%
Chemical Name or Material 1H,1H,10H,10H-Perfluoro-1,10-decanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.