accessibility menu, dialog, popup

UserName

CAS RN 7540-51-4

CH3(S)HOCH3CH3

CAS RN 7540-51-4

IUPAC Name: (3S)-3,7-dimethyloct-6-en-1-ol
Synonyms: (-)-citronellol l-citronellol (S)-(-)-citronellol (-)-3,7-dimethyloct-6-en-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: QMVPMAAFGQKVCJ-JTQLQIEISA-N
SMILES: C[C@H](CCO)CCC=C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results