missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Citronellol, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | L-Citronellol |
| CAS | 7540-51-4 |
| Molecular Formula | C10 H20 O |
| Quantity | 100 g |
| Synonym | 6-Octen-1-ol, 3,7-dimethyl-, (-)- (8CI), 6-Octen-1-ol, 3,7-dimethyl-, (S)-, (3S)-3,7-Dimethyl-6-octen-1-ol, (-)-(S)-Citronellol, (-)-Citronellol, (-)-β-Citronellol, (3S)-3,7-Dimethyloct-6-en-1-ol, (S)-(-)-Citronellol, (S)-(-)-β-Citronellol, (S)-3,7-Dimethyl-6-octen-1-ol, (S)-Citronellol, (S)-β-Citronellol, L-Citronellol, l-Citronellol, (3S)-3,7-Dimethyl-6-octen-1-ol |
| InChI Formula | InChI=1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1 |
| SMILES | C[C@H](CCO)CCC=C(C)C |
| IUPAC Name | (3S)-3,7-dimethyloct-6-en-1-ol |
| Molecular Weight (g/mol) | 156.27 |
| Formula Weight | 156.1514 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction