accessibility menu, dialog, popup

UserName

FFFClFFF

CAS RN 75462-59-8

IUPAC Name: 1-(chloromethyl)-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-(chloromethyl)-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 262.58
Molecular Formula: C9H5ClF6
InChi Key: OINTXXMBRBLMHH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(CCl)=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results