accessibility menu, dialog, popup

UserName

H3COOOH3COOHCH3HO(S)CH3O

CAS RN 7562-61-0

IUPAC Name: (2S)-4,10-diacetyl-11,13-dihydroxy-2,12-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(13),6,9,11-tetraene-3,5-dione
Synonyms: (2S)-4,10-diacetyl-11,13-dihydroxy-2,12-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(13),6,9,11-tetraene-3,5-dione
Molecular Weight (g/mol): 344.32
Molecular Formula: C18H16O7
InChi Key: CUCUKLJLRRAKFN-KKIBXBACSA-N
SMILES: CC(=O)C1C(=O)C=C2OC3=C(C(C)=O)C(O)=C(C)C(O)=C3[C@]2(C)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results