accessibility menu, dialog, popup

UserName

CAS RN 75822-54-7

OOBrHNO

CAS RN 75822-54-7

IUPAC Name: 5'-bromo-1',2'-dihydrospiro[1,3-dioxolane-2,3'-indol]-2'-one
Synonyms: 5'-bromo-1'H-spiro[1,3-dioxolane-2,3'-indol]-2'-one
Molecular Weight (g/mol): 270.08
Molecular Formula: C10H8BrNO3
InChi Key: GQAJPVMAVMEQQJ-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC(=O)C22OCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results