missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one, 99%, Thermo Scientific™

Product Code. 11420060
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11420060 1 g 1g
11410060 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 11420060

Brand: Thermo Scientific Alfa Aesar H50582.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75822-54-7
Molecular Formula C10H8BrNO3
Molecular Weight (g/mol) 270.08
MDL Number MFCD00097597
InChI Key GQAJPVMAVMEQQJ-UHFFFAOYSA-N
Synonym 5'-bromospiro 1,3-dioxolane-2,3'-indol-2' 1'h-one, 5'-bromospiro 1,3 dioxolane-2,3'-indolin-2'-one, 5'-bromo-1',2'-dihydrospiro 1,3-dioxolane-2,3'-indole-2'-one, 10-bromospiro 1,3-dioxolane-2,3'-indoline-7-one, 5'-bromospiro 1,3-dioxolane-2,3'-1h-indole-2'-one, maybridge1_007236, chembl6720, 5-bromoisatin 3-ethylene acetal, 5-bromospiro indoline-3,2'-1,3 dioxolane-2-one, 5/'-bromospiro 1,3-dioxolane-2,3/'-1h-indole-2/'-one
PubChem CID 2732922
IUPAC Name 5'-bromospiro[1,3-dioxolane-2,3'-1H-indole]-2'-one
SMILES BrC1=CC2=C(NC(=O)C22OCCO2)C=C1

Specifications

Quantity 1 g
Formula Weight 270.08
Percent Purity 99%
Chemical Name or Material 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.